Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDHA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597300 Supplier Invitrogen™ Supplier No. PA597300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HGVCYTRGLS VRSILAELTG RRGGCAKGKG GSMHMYTKNF YGGNGIVGAQ GPLGAGIALA CKYKGNDEIC LTLYGDGAAN QGQIAEAFNM AALWKLPCVF ICENNLYGMG TSTERAAASP DYYKRGNFIP GLKVDGMDVL CVREATKFAA NYCRSGKGPI LMELQTYRYH GHSMSDPGVS YRTREEIQEV RSKRDPIIIL QDRMVNSKLA TVEELKEIGA EVRKEIDDAA QFATTDPEPH LEELGHHIYS SDSSFEVRGA NPWIKFKSVS; Positive Samples: NCI-H460, SKOV3, A549, Mouse testis, Mouse ovary; Cellular Location: Mitochondrion matrix.

PDHA2 (Pyruvate Dehydrogenase E1 Alpha 2 Subunit) is a Protein Coding gene. Diseases associated with PDHA2 include Spermatogenic Failure 1. Among its related pathways are Citrate cycle (TCA cycle)and Central carbon metabolism in cancer. Gene Ontology (GO) annotations related to this gene include oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptorandpyruvate dehydrogenase (acetyl-transferring) activity. An important paralog of this gene isPDHA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDHA2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.886 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PDHA2
Gene Accession No. P29803, P35487, Q06437
Gene Alias Pdha2; Pdha-2; Pdhal; PDHE1-A type II; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 2; pyruvate dehydrogenase E1 alpha 2; PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT, TESTIS-SPECIFIC FORM, MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II); pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific
Gene Symbols PDHA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 119-388 of human PDHA2 (NP_0053811).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117098, 18598, 5161
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.