Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDHA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106454 Supplier Invitrogen™ Supplier No. PA5106454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PDHA2.

PDHA2 (Pyruvate Dehydrogenase E1 Alpha 2 Subunit) is a Protein Coding gene. Diseases associated with PDHA2 include Spermatogenic Failure 1. Among its related pathways are Citrate cycle (TCA cycle)and Central carbon metabolism in cancer. Gene Ontology (GO) annotations related to this gene include oxidoreductase activity, acting on the aldehyde or oxo group of donors, disulfide as acceptorandpyruvate dehydrogenase (acetyl-transferring) activity. An important paralog of this gene isPDHA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDHA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PDHA2
Gene Accession No. P29803, P35487, Q06437
Gene Alias Pdha2; Pdha-2; Pdhal; PDHE1-A type II; pyruvate dehydrogenase; pyruvate dehydrogenase (lipoamide) alpha 2; pyruvate dehydrogenase E1 alpha 2; PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT, TESTIS-SPECIFIC FORM, MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II); pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific
Gene Symbols PDHA2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PDHA2(Accession P29803), corresponding to amino acid residues L333-K383.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117098, 18598, 5161
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.