Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDHX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PDHX |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PDHX Polyclonal specifically detects PDHX in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PDHX | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenasecomplex, DLDBP, E3-binding protein, E3BPpyruvate dehydrogenase complex, E3-binding protein subunit, OPDX, PDX1Lipoyl-containing pyruvate dehydrogenase complex component X, proXpyruvate dehydrogenase complex, lipoyl-containing component X, pyruvate dehydrogenase complex, component X, pyruvate dehydrogenase protein X component, mitochondrial | |
PDHX | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
8050 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PAARNILEKHSLDASQGTATGPRGIFTKEDALKLVQLKQTGKITESRPTPAPTATPTAPSPLQATAGPSYPRPVIPPVSTPGQPNAVGTFTEI | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title