Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDHX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583422
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583422 Supplier Invitrogen™ Supplier No. PA583422
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAARNILEKH SLDASQGTAT GPRGIFTKED ALKLVQLKQT GKITESRPTP APTATPTAPS PLQATAGPSY PRPVIPPVST PGQPNAVGTF TEI Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 80%.

The PDHX gene encodes component X of the pyruvate dehydrogenase (PDH) complex. For a detailed description of the pyruvate dehydrogenase complex. The mammalian PDH complex differs from that in E. coli and from the other mammalian alpha-keto acid dehydrogenases by the presence of a 53-kDa protein called protein X. Component X binds to the E3 component of the PDH complex (Robinson et al., 1990 ; Aral et al., 1997 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDHX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pdhx
Gene Accession No. O00330
Gene Alias Ac1164; AI481367; component X; Dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex; dihydrolipoamide dehydrogenase-binding protein of pyruvate dehydrogenase complex (E3-binding protein) (proX); DLDBP; E3-binding protein; E3BP; GSF; IDX-1; IPF1; IPF-1; IUF-1; lipoyl-containin; lipoyl-containing pyruvate dehydrogenase complex component X; OPDX; Pdhx; PDX1; PDX-1; proX; pyruvate dehydrogenase; pyruvate dehydrogenase complex component X; pyruvate dehydrogenase complex, component X; pyruvate dehydrogenase complex, E3-binding protein subunit; pyruvate dehydrogenase complex, lipoyl-containing component X; pyruvate dehydrogenase protein X component, mitochondrial; STF-1
Gene Symbols Pdhx
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PDHX. Recombinant protein control fragment (Product #RP-97305).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.