Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PDI, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABS1655 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1655 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1655 Supplier MilliporeSigma™ Supplier No. ABS1655

Rabbit Polyclonal Antibody

Protein disulfide-isomerase (EC 5.3.4.1; UniProt P07237; also known as Cellular thyroid hormone-binding protein, p55, P4Hbeta, PDI, Prolyl 4-hydroxylase subunit beta) is encoded by the P4HB (also known as CLCRP1, ERBA2L, PDI, PDIA1, PO4DB) gene (Gene ID 5034) in human. Protein disulphide isomerase (PDI), BiP (GRP78 or heat shock 70kDa protein 5), and calreticulin (CRT) are ER lumen folding factors that, together with other PTM enzymes, assist the process of newly synthesized proteins before their release from ER. PDI, BiP, and CRT themselves are subject to posttranslational signal peptide removal after their synthesis, exposing D18, E19, and E18 at the newly formed N-terminus, respectively. The N-terminus D and E residues of the mature proteins are permissive to arginine-tRNA-protein transferase 1/ATE1-catalyzed arginylation, forming arginylated mature proteins (R-DPI, R-BiP, and R-CRT) with altered cellular localization to allow their participation in non-ER functions. Although only the basal levels of R-PDI and R-CRT, but not R-BiP, are generally detectible in non-stimulated cells, upregulated N-terminal arginylation of all three proteins is observed upon cytosolic dsDNA exposure and proteasomal inhibition, indicating a shared role in innate immune responses to invading microbes. In addition, ER stress induction by thapsigargin treatment synergistically boosts proteasomal inhibition-induced upregulation of cellular levels of R-DPI, R-BiP, and R-CRT, suggesting that ER stress accelerates the supply of ER lumenal DPI, BiP, and CRT for N-terminal arginylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDI
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.05% Sodium Azide.
Gene P4HB, CLCRP1, ERBA2L, PDI, PDIA1, PO4DB
Gene Accession No. P07237
Host Species Rabbit
Immunogen Linear peptide corresponding to the N-terminal sequence of arginylated mature human PDI.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000909
Test Specificity This rabbit polyclonal antibody specifically detected the immunogen peptide, but not the control peptide without arginylation at the N-terminal Asp18 (Cha-Molstad, H., et al. (2015). Nat. Cell Biol. 17(7):917-929).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.