Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDIA2 Antibody (SR2029), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32170950UL
Description
PDIA2 Monoclonal antibody specifically detects PDIA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
PDIA2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR2029 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
Pancreas-specific protein disulfide isomerase, pancreatic protein disulfide isomerase, PDA2, PDI, PDIp, PDIR, protein disulfide isomerase, pancreatic, protein disulfide isomerase-associated 2, protein disulfide-isomerase A2, Rho GDP dissociation inhibitor gamma | |
A synthesized peptide derived from human PDIA2 (Uniprot #: Q13087) | |
50 μg | |
Biologically Active Proteins, Cancer, Cellular Markers, ER Markers, Hypoxia | |
64714 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction