Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDIA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588756 Supplier Invitrogen™ Supplier No. PA588756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FGLKAEAAPT LRLVNLETTK KYAPVDGGPV TAASITAFCH AVLNGQVKPY LLSQEIPPDW DQRPVKTLVG KNFEQVAFDE TKNVFVKFYA PWCTHCKEMA PAWEALAEKY QDHEDIIIAE LDATANELDA FAVHGFPTLK YFPAGPGRKV IEYKSTRDLE TFSKFLDNGG VLPTEEPPEE PAAPFPEPPA NSTMGSKEEL; Positive Samples: U-87MG, Mouse liver, Rat liver; Cellular Location: Endoplasmic reticulum lumen.

Protein disulfide isomerases resident proteins that catalyze protein folding and thiol-disulfide interchange reactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDIA2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PDIA2
Gene Accession No. D3Z6P0, Q13087
Gene Alias 1810041F13Rik; AI661267; GIT antibody; LA16c-314G4.2; Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2; PDI; Pdia2; Pdip; Pdipl; PDIR; protein disulfide isomerase (pancreas) like; protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A member 2; protein disulfide isomerase family A, member 2; protein disulfide isomerase, pancreatic; protein disulfide isomerase-associated 2; protein disulfide-isomerase A2; Rho GDP dissociation inhibitor gamma
Gene Symbols PDIA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 326-525 of human PDIA2 (NP_0068402).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287164, 64714, 69191
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.