Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDIA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117368
Description
Centrifuge briefly prior to opening.
Protein disulfide isomerases resident proteins that catalyze protein folding and thiol-disulfide interchange reactions.
Specifications
| PDIA2 | |
| Polyclonal | |
| Unconjugated | |
| PDIA2 | |
| 1810041F13Rik; AI661267; GIT antibody; LA16c-314G4.2; Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2; PDI; Pdia2; Pdip; Pdipl; PDIR; protein disulfide isomerase (pancreas) like; protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A member 2; protein disulfide isomerase family A, member 2; protein disulfide isomerase, pancreatic; protein disulfide isomerase-associated 2; protein disulfide-isomerase A2; Rho GDP dissociation inhibitor gamma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64714 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.51 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q13087 | |
| PDIA2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human PDIA2 / PDIP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction