Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDIA2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518690
Description
This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Protein disulfide isomerases resident proteins that catalyze protein folding and thiol-disulfide interchange reactions.
Specifications
| PDIA2 | |
| Polyclonal | |
| Unconjugated | |
| PDIA2 | |
| 1810041F13Rik; AI661267; GIT antibody; LA16c-314G4.2; Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2; PDI; Pdia2; Pdip; Pdipl; PDIR; protein disulfide isomerase (pancreas) like; protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A member 2; protein disulfide isomerase family A, member 2; protein disulfide isomerase, pancreatic; protein disulfide isomerase-associated 2; protein disulfide-isomerase A2; Rho GDP dissociation inhibitor gamma | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 64714 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q13087 | |
| PDIA2 | |
| Synthetic peptide sequence (NLETTKKYAPVD) corresponding to the internal amino acids of PDIA2 (aa 340-352). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction