Learn More
PDIA2 Rabbit anti-Human, Polyclonal, Bioss

Description
Specifications
Specifications
| Antigen | PDIA2 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH |
| Gene | PDIA2 |
| Gene Accession No. | Q13087 |
| Gene Alias | 1810041F13Rik; AI661267; GIT antibody; LA16c-314G4.2; Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2; PDI; Pdia2; Pdip; Pdipl; PDIR; protein disulfide isomerase (pancreas) like; protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A member 2; protein disulfide isomerase family A, member 2; protein disulfide isomerase, pancreatic; protein disulfide isomerase-associated 2; Protein disulfide-isomerase A2; Rho GDP dissociation inhibitor gamma |
| Gene Symbols | PDIA2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.