Learn More
PDIA3 Mouse anti-Human, Clone: OTI2D2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504993

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates.Specifications
PDIA3 | |
Monoclonal | |
Unconjugated | |
PDIA3 | |
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC | |
Mouse | |
Affinity Chromatography | |
RUO | |
2923 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P30101 | |
PDIA3 | |
Human recombit protein fragment corresponding to amino acids 140-505 of human PDIA3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.