Learn More
PDIA3 Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human PDIA3.
Supplier: Abnova Corporation PAB29388
Description
This gene encodes a protein of the endoplasmic reticulum that interacts with lectin chaperones calreticulin and calnexin to modulate folding of newly synthesized glycoproteins. The protein was once thought to be a phospholipase; however, it has been demonstrated that the protein actually has protein disulfide isomerase activity. It is thought that complexes of lectins and this protein mediate protein folding by promoting formation of disulfide bonds in their glycoprotein substrates. [provided by RefSeq]
Sequence: PTLKIFRDGEEAGAYDGPRTADGIVSHLKKQAGPASVPLRTEEEFKKFISDKDASIVGFFDDSFSEAHSEFLKAASNLRDNYRFAHTNVESLVNEYDDNGEGIILFRPSHLTNKFEDKSpecifications
PDIA3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant human PDIA3. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
PDIA3 | |
Recombinant protein corresponding to human PDIA3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:2500-1:5000) Immunofluorescence (1-4 ug/mL) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
ER60/ERp57/ERp60/ERp61/GRP57/GRP58/HsT17083/P58/PI-PLC | |
Rabbit | |
Antigen affinity purification | |
RUO | |
2923 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.