Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDIA4 Monoclonal Antibody (OTI6B6), TrueMAB™, OriGene
SDP

Catalog No. 502540653
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0653 30 μL
50-167-4670 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0653 Supplier OriGene Supplier No. TA503888S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDIA4 Monoclonal antibody specifically detects PDIA4 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PDIA4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI6B6
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDIA4
Gene Accession No. P13667
Gene Alias AI987846; caBP2; Cai; calcium binding protein, intestinal; calcium-binding protein 2; EC 5.3.4.1; endoplasmic reticulum protein 72; endoplasmic reticulum resident protein 70; Endoplasmic reticulum resident protein 72; ER protein 70; ER protein 72; Erp70; Erp72; ERp-72; PDIA4; protein disu; protein disulfide isomerase A4; protein disulfide isomerase associated 4; protein disulfide isomerase family A member 4; protein disulfide isomerase family A, member 4; protein disulfide isomerase related protein (calcium-binding protein, intestinal-related); protein disulfide isomerase-associated 4; protein disulfide-isomerase A4; Protein ERp-72; testicular tissue protein Li 137
Host Species Mouse
Immunogen Full length human recombinant protein of human PDIA4 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9601
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.