Learn More
PDIA4 Mouse anti-Human, Clone: OTI2E11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503886

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.Specifications
PDIA4 | |
Monoclonal | |
Unconjugated | |
PDIA4 | |
ERP70, ERP72, ERp-72 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9601 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2E11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P13667 | |
PDIA4 | |
Full length human recombit protein of human PDIA4 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.