Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDIA6 Mouse anti-Human, Mouse, Rat, Clone: 1G2D4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | PDIA6 |
---|---|
Clone | 1G2D4 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7086
![]() ![]() |
Proteintech Group Inc
666691IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to PDIA6.
P5 is a novel protein that was originally identified in humans. Functionally, P5 has been shown to have peptide binding ability and chaperone activity specific to certain proteins. P5 shares several structural similarities to PDI, an important protein of the ER. The deduced amino acid sequence of P5 shows that it contains the ER retention signal KDEL. P5 also contains two CXXC-like motifs, a motif responsible for oxidoreductase activity. These two CXXC motifs are found on the N- and C-terminus of the protein, one at each end. Studies suggest that the N-terminal CXXC motif is more important than the second for isomerase activity. Contrary to most of the PDI-like proteins, P5 is not stress-inducible.Specifications
PDIA6 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ER protein 5, ERP5, P5, PDIA6, Protein disulfide isomerase A6, Protein disulfide isomerase P5, TXNDC7 | |
Pdia6 | |
IgG1 | |
Protein G | |
Antibody |
1G2D4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q15084, Q63081, Q922R8 | |
10130, 286906, 71853 | |
PDIA6 Fusion Protein Ag27608 | |
Primary | |
-20°C | |
PDIA6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title