Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDIA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303564100UL
Description
PDIA6 Polyclonal specifically detects PDIA6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
PDIA6 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
10130 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown | |
Unconjugated | |
EC 5.3.4.1, Endoplasmic reticulum protein 5, ERp5, ERP5ER protein 5, P5TXNDC7, protein disulfide isomerase family A, member 6, Protein disulfide isomerase P5, protein disulfide isomerase-associated 6, protein disulfide isomerase-related protein, protein disulfide-isomerase A6, thioredoxin domain containing 7 (protein disulfide isomerase), Thioredoxin domain-containing protein 7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-300 of human PDIA6 (NP_005733.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction