Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDIK1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101862 Supplier Invitrogen™ Supplier No. PA5101862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PDIK1L.

PDIK1L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDIK1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PDIK1L
Gene Accession No. Q8N165, Q8QZR7
Gene Alias BC027088; casein kinase; CLIK1L; PDIK1L; PDLIM1 interacting kinase 1 like; PDLIM1-interacting kinase 1-like; Serine/threonine-protein kinase PDIK1L; STK35L2
Gene Symbols PDIK1L
Host Species Rabbit
Immunogen A synthesized peptide derived from human PDIK1L(Accession Q8N165), corresponding to amino acid residues V118-D168.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149420, 230809
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.