Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDILT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113129
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113129 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113129 Supplier Invitrogen™ Supplier No. PA5113129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDILT Polyclonal Antibody for Western Blot, ELISA

Probable redox-inactive chaperone involved in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDILT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PDILT
Gene Accession No. Q8N807
Gene Alias PDIA7; PDILT; protein disulfide isomerase family A, member 7; protein disulfide isomerase like, testis expressed; protein disulfide isomerase-like protein of the testis; protein disulfide isomerase-like, testis expressed; protein disulfide-isomerase-like protein of the testis
Gene Symbols PDILT
Host Species Rabbit
Immunogen Recombinant Human Protein disulfide-isomerase-like protein of the testis protein (494-584AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 204474
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.