Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDIR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522384
Description
Recommended positive controls: A549, HepG2, HCT116. Predicted reactivity: Mouse (90%), Rat (91%), Sheep (91%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PDIR (protein disulfide isomerase-related protein) is a novel PDI protein that was originally isolated from a human placental cDNA library. It has been suggested that from its nucleotide sequence, PDIR has three CXXC-like motifs (Cys-Ser-Met-Cys, Cys-Gly-His-Cys and Cys-Pro-His-Cys), which are found in the PDI superfamily of proteins. The CXXC motif is responsible for oxidoreductase activity. PDIR is also shown to have a putative endoplasmic reticulum (ER) retention signal 'Lys-Glu-Glu-Leu' at its carboxyl terminus, indicative of ER resident proteins. Northern blot analysis of PDIR indicates that it is preferentially expressed in cells actively secreting proteins. It has also been shown that expression of PDIR is stress-inducible. Data suggest that PDIR, like other PDIs, has oxidoreductase activity of disulfide bonds and facilitates the protein folding process within the lumen of the ER.
Specifications
| PDIR | |
| Polyclonal | |
| Unconjugated | |
| PDIA5 | |
| 2700053F16Rik; AU015525; Pdia5; PDIR; protein disulfide isomerase A5; protein disulfide isomerase associated 5; protein disulfide isomerase family A member 5; protein disulfide isomerase family A, member 5; protein disulfide isomerase-associated 5; protein disulfide isomerase-related protein; Protein disulfide-isomerase A5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10954, 72599 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.52 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14554, Q921X9 | |
| PDIA5 | |
| Recombinant protein encompassing a sequence within the center region of human PDIR. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction