Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDIR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PDIR |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0303
![]() ![]() |
Proteintech Group Inc
155451AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PDIR (protein disulfide isomerase-related protein) is a novel PDI protein that was originally isolated from a human placental cDNA library. It has been suggested that from its nucleotide sequence, PDIR has three CXXC-like motifs (Cys-Ser-Met-Cys, Cys-Gly-His-Cys and Cys-Pro-His-Cys), which are found in the PDI superfamily of proteins. The CXXC motif is responsible for oxidoreductase activity. PDIR is also shown to have a putative endoplasmic reticulum (ER) retention signal ′Lys-Glu-Glu-Leu′ at its carboxyl terminus, indicative of ER resident proteins. Northern blot analysis of PDIR indicates that it is preferentially expressed in cells actively secreting proteins. It has also been shown that expression of PDIR is stress-inducible. Data suggest that PDIR, like other PDIs, has oxidoreductase activity of disulfide bonds and facilitates the protein folding process within the lumen of the ER.Specifications
PDIR | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q14554, Q5I0H9, Q921X9 | |
10954, 360722, 72599 | |
PDIR Fusion Protein Ag7901 | |
Primary | |
-20°C | |
PDIA5 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PDIA5, PDIR, Protein disulfide isomerase A5 | |
Pdia5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title