Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDK-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP244132
Description
Specifically detects PDPK1 in C. elegans, Human samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting
PDK-1 Polyclonal specifically detects PDK-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| PDPK1 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| 3-phosphoinositide-dependent protein kinase 1, 3-phosphoinositide-dependent protein kinase 2 pseudogene, PDK1, PDPK2, PDPK2P, PkB kinase like gene 1 | |
| Rabbit | |
| 49.24 kDa | |
| 0.1 mg | |
| Cancer, mTOR Pathway, Signal Transduction | |
| 5170 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry | |
| 1.3 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:100000, Immunohistochemistry 1:10-1:500 | |
| Q15118 | |
| PDPK1 | |
| This affinity purified PDK-1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal of human PDK-1 protein. (Uniprot: Q15118) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction