Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDK-1, p Tyr9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323292
Description
PDK-1 Polyclonal antibody specifically detects PDK-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PDK-1 | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200 | |
3-phosphoinositide-dependent protein kinase 1, 3-phosphoinositide-dependent protein kinase 2 pseudogene, PDK1, PDPK2, PDPK2P, PkB kinase like gene 1 | |
Phospho-PDK-1 (Tyr-9) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding tyrosine 9 in human PDK-1. This sequence is highly conserved in rat and mouse PDK-1. [UniProt# O15530] | |
0.1 mL | |
Cancer, mTOR Pathway, Phospho Specific, Signal Transduction | |
5170 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction