Learn More
Promega PDK1 Kinase Enzyme System, 1mg
Catalog No. PRV2762 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2762 1 mg
1 options
Catalog No. PRV2762 Supplier Promega Supplier No. V2762
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PDK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. PDK1 (3-phosphoinositide-dependent protein kinase) is activated by the presence of PtdIns(3,4,5)P3 or PtdIns(3,4)P2.
  • Easily Screen and Profile PDK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C