Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDK1 Recombinant Rabbit Monoclonal Antibody (5T6S8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535736
Description
Immunogen sequence: STAPRPRVET SRAVPLAGFG YGLPISRLYA QYFQGDLKLY SLEGYGTDAV IYIKALSTDS IERLPVYNKA AWKHYNTNHE ADDWCVPSRE PKDMTTFRSA.
PDK1 (3-phosphoinositide dependent kinase-1) is a serine/threonine protein kinase localized to the plasma membrane. It plays a vital role in insulin-induced glycogen synthesis, PKC activation, and activation of the AGC subfamily of protein kinases. Plays an important role in cellular responses to hypoxia and is important for cell proliferation under hypoxia. Protects cells against apoptosis in response to hypoxia and oxidative stress.
Specifications
PDK1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q15118, Q63065, Q8BFP9 | |
PDK1 | |
A synthetic peptide corresponding to a sequence within amino acids 337-436 of human PDK1/PDHK1 (NP_0026011). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5T6S8 | |
Unconjugated | |
PDK1 | |
[Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 1, mitochondrial; B830012B01; D530020C15Rik; isoenzyme 1; kinase isoenzyme 1; lipoamide; mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1; PDH kinase 1; PDHK1; PDK p48; Pdk1; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 1; Pyruvate dehydrogenase kinase isoform 1; pyruvate dehydrogenase kinase, isoenzyme 1; pyruvate dehydrogenase kinase, isozyme 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
116551, 228026, 5163 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction