Learn More
PDK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 182621AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-2076
Only null left
Add to Cart
Description
Description
This antibody is specific to PDK1.
Serine/threonine kinase that plays a key role in regulation of glucose and fatty acid metabolism and homeostasis via phosphorylation of the pyruvate dehydrogenase subunits PDHA1 and PDHA2. This inhibits pyruvate dehydrogenase activity, and thereby regulates metabolite flux through the tricarboxylic acid cycle, down-regulates aerobic respiration and inhibits the formation of acetyl-coenzyme A from pyruvate. Plays an important role in cellular responses to hypoxia and is important for cell proliferation under hypoxia. Protects cells against apoptosis in response to hypoxia and oxidative stress.
Specifications
Specifications
PDK1
Polyclonal
Unconjugated
PDK1
PDH kinase 1, PDHK1, PDK1, Pyruvate dehydrogenase kinase isoform 1
Rabbit
Antigen Affinity Chromatography
RUO
116551, 228026, 5163
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15118, Q63065, Q8BFP9
PDK1
PDK1 Fusion Protein Ag13121
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.