Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540426
Description
Peptide sequence: ELFKNAMRAT VESHESSLIL PPIKVMVALG EEDLSIKMSD RGGGVPLRKI Sequence homology: Cow: 77%; Dog: 92%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Zebrafish: 77%.
PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
Specifications
PDK2 | |
Polyclonal | |
Unconjugated | |
PDK2 | |
[Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 2, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 2, mitochondrial; pyruvate dehydrogenase kinase, isozyme 2; LOC524075 protein; PDH kinase 2; PDHK2; PDK P45; PDK2; PDKII; pyruvate dehydrogenase; pyruvate dehydrogenase 2; pyruvate dehydrogenase kinase 2; pyruvate dehydrogenase kinase 2 subunit p45 (PDK2); Pyruvate dehydrogenase kinase isoform 2; pyruvate dehydrogenase kinase, isoenzyme 2; pyruvate dehydrogenase kinase, isozyme 2; pyruvate dehydrogenase, lipoamide, kinase isozyme 2, mitochondrial | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5164 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15119 | |
PDK2 | |
Synthetic peptide directed towards the middle region of human PDK2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction