Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120166 Supplier Invitrogen™ Supplier No. PA5120166
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SH-SY5Y. The target is usually found in the following locations: Mitochondrion matrix. Immunogen sequence: FGFDPFISTN IQYFLDRFYT NRISFRMLIN QHTLLFGGDT NPVHPKHIGS IDPTCNVADV VKDAYETAKM LCEQYYLVAP ELEVEEFNAK APDKPIQVVY VPSHLFHMLF ELFKNSMRAT VELYEDRKEG YPAVKTLVTL GKEDLSIKIS DLGGGVPLRK IDRLFNYMYS TAPRPSLEPT RAAPLAGFGY GLPISRLYAR YFQGDLKLYS MEGVGTDAVI YLKALSSESF ERLPVFNKSA WRHYKTTPEA DDWSNPSSEP RDASKYKAKQ.

PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDK3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PDK3
Gene Accession No. Q15120
Gene Alias [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial; 2610001M10Rik; AI035637; CMTX6; GS1-358P8.4; PDHK3; PDK3; pyruvate dehydrogenase; Pyruvate dehydrogenase kinase 3; Pyruvate dehydrogenase kinase isoform 3; pyruvate dehydrogenase kinase, isoenzyme 3; pyruvate dehydrogenase kinase, isozyme 3; pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial
Gene Symbols PDK3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 137-406 of human PDK3 (NP_005382.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5165
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.