Learn More
PDK4 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals NBP107049AF647
Catalog No. NBP107049Y
Only null left
Add to Cart
Description
Description
PDK4 Polyclonal specifically detects PDK4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PDK4
Polyclonal
Western Blot, Immunocytochemistry/Immunofluorescence
Q16654
PDK4
Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 200-250). [Swiss-Prot# Q16654]
Affinity Purified
RUO
Primary
Human. Immunogen sequence has 94% homology with rat and 88% homology with mouse.
Store at 4C in the dark.
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
[Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial
Rabbit
49 kDa
0.1 mL
Cancer, Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction
5166
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only