Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDK4 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP107049UV
Description
PDK4 Polyclonal specifically detects PDK4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PDK4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction | |
| 5166.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PDK4 | |
| Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 200-250). [Swiss-ProtNo. Q16654] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction