Learn More
Invitrogen™ PDK4 Polyclonal Antibody

Description
Immunogen sequence homology: rat (94%), bovine (100%). Not recommended for ICC, as nonspecific nuclear staining was observed in HeLa cells.
Specifications
Specifications
| Antigen | PDK4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | Pdk4 |
| Gene Accession No. | O70571, Q16654 |
| Gene Alias | [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 4, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial; AV005916; kinase isozyme 4; lipoamide; PDHK4; Pdk4; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 4; Pyruvate dehydrogenase kinase isoform 4; pyruvate dehydrogenase kinase, isoenzyme 4; pyruvate dehydrogenase kinase, isozyme 4; pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial; pyruvate dehydrogenate kinase 4 |
| Gene Symbols | Pdk4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.