Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298546MFV450
Description
PDLIM1 Polyclonal antibody specifically detects PDLIM1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PDLIM1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1 | |
Produced in rabbits immunized with E. coli-derived Human PDLIM1 fragment. | |
0.1 mL | |
Cancer, Endocrinology | |
9124 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction