Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP151917
Description
PDLIM1 Polyclonal specifically detects PDLIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
PDLIM1 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:64000 | |
NP_066272.1 | |
PDLIM1 | |
Peptide with sequence C-TPPEGYEVVTVFPK corresponding to C-Terminus according to NP_066272.1. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
9124 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction