Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM1 Antibody (SR1603), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PDLIM1 |
---|---|
Clone | SR1603 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
PDLIM1 Monoclonal antibody specifically detects PDLIM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
PDLIM1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1 | |
A synthesized peptide derived from human PDLIM1 (Uniprot #: O00151) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1603 | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
9124 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title