Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDLIM1 Recombinant Rabbit Monoclonal Antibody (JE55-21)
GREENER_CHOICE

Catalog No. PIMA536211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536211 Supplier Invitrogen™ Supplier No. MA536211
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

PDLIM1 Recombinant Monoclonal Antibody for Flow, IHC (P), Western Blot

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. A distinctive feature of TBP is a long string of glutamines in the N-terminal. This region of the protein modulates the DNA binding activity of the C terminus, and modulation of DNA binding affects the rate of transcription complex formation and initiation of transcription. Mutations that expand the number of CAG repeats encoding this polyglutamine tract, and thus increase the length of the polyglutamine string, are associated with spinocerebellar ataxia 17, a neurodegenerative disorder classified as a polyglutamine disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDLIM1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE55-21
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Pdlim1
Gene Accession No. O00151, O70400, P52944
Gene Alias carboxyl terminal LIM domain protein 1; Clim1; Clp36; CLP-36; C-terminal LIM domain protein 1; elfin; enigma; epididymis secretory protein Li 112; hCLIM1; HEL-S-112; LIM domain protein CLP-36; LIM protein; mClim1; Pdlim1; PDZ and LIM domain 1; PDZ and LIM domain 1 (elfin); PDZ and LIM domain protein 1
Gene Symbols Pdlim1
Host Species Rabbit
Immunogen Recombinant protein within Human PDLIM1 aa 181-285.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54132, 54133, 9124
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.