Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM2 Antibody (OTI11A12), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273311MFV450
Description
PDLIM2 Monoclonal antibody specifically detects PDLIM2 in Human samples. It is validated for Flow CytometrySpecifications
PDLIM2 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
64236 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI11A12 | |
Flow Cytometry | |
FLJ34715, MYSTIQUE, PDZ and LIM domain 2 (mystique), PDZ and LIM domain protein 2, PDZ-LIM protein mystique, SLIM | |
Full length human recombinant protein of human PDLIM2 (NP_067643) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction