Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM2 Antibody (OTI11A12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200619
Description
PDLIM2 Monoclonal specifically detects PDLIM2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PDLIM2 | |
| Monoclonal | |
| 0.62 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:100 | |
| PDLIM2 | |
| Full length human recombinant protein of human PDLIM2 (NP_067643) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 64236 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI11A12 | |
| Unconjugated | |
| FLJ34715, MYSTIQUE, PDZ and LIM domain 2 (mystique), PDZ and LIM domain protein 2, PDZ-LIM protein mystique, SLIM | |
| Mouse | |
| 37.3 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction