Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM2 Monoclonal Antibody (OTI1D12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502709S


Description
PDLIM2 Monoclonal antibody specifically detects PDLIM2 in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
PDLIM2 | |
Monoclonal | |
0.68 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96JY6 | |
Mouse | |
Affinity chromatography | |
RUO | |
486123, 64236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1D12 | |
Unconjugated | |
PDLIM2 | |
4732462F18Rik; hypothetical protein; mystique; Pdlim2; PDZ and LIM domain 2; PDZ and LIM domain 2 (mystique); PDZ and LIM domain protein 2; PDZ-LIM protein mystique; PP6345; Slim; STAT-interacting LIM protein | |
Full length human recombinant protein of human PDLIM2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction