Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM2 Monoclonal Antibody (OTI9B6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502711

Description
Probable adapter protein located at the actin cytoskeleton that promotes cell attachment. Necessary for the migratory capacity of epithelial cells. Overexpression enhances cell adhesion to collagen and fibronectin and suppresses anchorage independent growth. May contribute to tumor cell migratory capacity.Specifications
PDLIM2 | |
Monoclonal | |
0.41 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96JY6 | |
PDLIM2 | |
Full length human recombit protein of human PDLIM2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI9B6 | |
Unconjugated | |
PDLIM2 | |
MYSTIQUE, SLIM | |
Mouse | |
Affinity Chromatography | |
RUO | |
486123, 64236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction