Learn More
PDLIM2 Mouse anti-Canine, Human, Clone: OTI10C4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502708

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Probable adapter protein located at the actin cytoskeleton that promotes cell attachment. Necessary for the migratory capacity of epithelial cells. Overexpression enhances cell adhesion to collagen and fibronectin and suppresses anchorage independent growth. May contribute to tumor cell migratory capacity.Specifications
PDLIM2 | |
Monoclonal | |
Unconjugated | |
PDLIM2 | |
MYSTIQUE, SLIM | |
Mouse | |
Affinity Chromatography | |
RUO | |
486123, 64236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI10C4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96JY6 | |
PDLIM2 | |
Full length human recombit protein of human PDLIM2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.