Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDLIM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531484
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531484 Supplier Invitrogen™ Supplier No. PA531484
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Probable adapter protein located at the actin cytoskeleton that promotes cell attachment. Necessary for the migratory capacity of epithelial cells. Overexpression enhances cell adhesion to collagen and fibronectin and suppresses anchorage independent growth. May contribute to tumor cell migratory capacity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDLIM2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PDLIM2
Gene Accession No. Q96JY6
Gene Alias 4732462F18Rik; hypothetical protein; mystique; Pdlim2; PDZ and LIM domain 2; PDZ and LIM domain 2 (mystique); PDZ and LIM domain protein 2; PDZ-LIM protein mystique; PP6345; Slim; STAT-interacting LIM protein
Gene Symbols PDLIM2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PDLIM2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.