Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219755
Description
PDLIM3 Polyclonal specifically detects PDLIM3 in Human, Mouse samples. It is validated for Western Blot.Specifications
PDLIM3 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
PDLIM3 | |
Recombinant protein encompassing a sequence within the center region of human PDLIM3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
27295 | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:20000 | |
Actinin-associated LIM protein, ALPFLJ26096, Alpha-actinin-2-associated LIM protein, DKFZp686L0362, enigma homolog, PDZ and LIM domain 3, PDZ and LIM domain protein 3 | |
Rabbit | |
39 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction