Learn More
Invitrogen™ PDLIM3 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ALP.
Specifications
Specifications
| Antigen | PDLIM3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PDLIM3 |
| Gene Accession No. | O70209, Q53GG5, Q66HS7 |
| Gene Alias | actinin alpha 2 associated LIM protein; actinin-associated LIM protein; Actn2lp; AI463105; ALP; alpha-actinin-2-associated LIM protein; enigma homolog; PDLIM3; PDZ and LIM domain 3; PDZ and LIM domain protein 3; SK-2 |
| Gene Symbols | PDLIM3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.