Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PDLIM4 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518242
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes a protein which may be involved in bone development. Mutations in this gene are associated with susceptibility to osteoporosis.
Specifications
| PDLIM4 | |
| Polyclonal | |
| Unconjugated | |
| PDLIM4 | |
| enigma homolog; H-REV18; LIM domain protein; LIM protein RIL; Pdlim4; PDZ and LIM domain 4; PDZ and LIM domain protein 4; reversion induced LIM; reversion-induced LIM protein; RIL; RIT-18 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 8572 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P50479 | |
| PDLIM4 | |
| Synthetic peptide sequence (DVVAVYPNAKVELV) corresponding to the C-terminus amino acids of PDLIM4 (aa 317-330). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction