Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6093R

Description
This gene encodes a protein which may be involved in bone development. Mutations in this gene are associated with susceptibility to osteoporosis.Specifications
PDLIM4 | |
Polyclonal | |
Unconjugated | |
PDLIM4 | |
enigma homolog; H-REV18; LIM domain protein; LIM protein RIL; PDLIM4; PDZ and LIM domain 4; PDZ and LIM domain protein 4; reversion induced LIM; Reversion-induced LIM protein; RIL; RIT-18 | |
Rabbit | |
Protein A | |
RUO | |
8572 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P50479 | |
PDLIM4 | |
KLH conjugated synthetic peptide derived from human RIL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction