Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM5 Antibody (OTI1B5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273316APC
Description
PDLIM5 Monoclonal antibody specifically detects PDLIM5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PDLIM5 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
10611 | |
Store at 4C in the dark. | |
IgG2b |
Western Blot | |
OTI1B5 | |
Western Blot | |
Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5 | |
Full length human recombinant protein of human PDLIM5(NP_006448) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction