Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM5 Monoclonal Antibody (OTI1C11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504505

Description
May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.Specifications
PDLIM5 | |
Monoclonal | |
0.94 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96HC4 | |
PDLIM5 | |
Full length human recombit protein of human PDLIM5 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI1C11 | |
Unconjugated | |
PDLIM5 | |
ENH, ENH1, L9, LIM | |
Mouse | |
Affinity Chromatography | |
RUO | |
10611 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction