Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM5 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504448S


Description
PDLIM5 Monoclonal antibody specifically detects PDLIM5 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
PDLIM5 | |
Monoclonal | |
3.24 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O95628 | |
Mouse | |
Affinity chromatography | |
RUO | |
4850 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Western Blot | |
OTI1H10 | |
Unconjugated | |
CNOT4 | |
1110001A05Rik; AI987914; C87059; ENH; Enh1; Enh2; Enh3; Enigma homolog; enigma-like LIM domain protein; enigma-like PDZ and LIM domains protein; L9; LIM; Pdlim5; PDZ and LIM domain 5; PDZ and LIM domain protein 5 | |
Human recombinant protein fragment corresponding to amino acids 190-455 of human CNOT4 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction