Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM5 Antibody (OTI1H10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273317B
Description
PDLIM5 Monoclonal specifically detects PDLIM5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
| PDLIM5 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PDLIM5 | |
| Full length recombinant protein of human PDLIM5 (NP_006448) produced in HEK293T cells. | |
| 0.1 mL | |
| 10611.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI1H10 | |
| Western Blot, Flow Cytometry | |
| Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction