Learn More
PDLIM5 Mouse anti-Human, Clone: OTI1C11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504505

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.Specifications
PDLIM5 | |
Monoclonal | |
Unconjugated | |
PDLIM5 | |
ENH, ENH1, L9, LIM | |
Mouse | |
Affinity Chromatography | |
RUO | |
10611 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1C11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96HC4 | |
PDLIM5 | |
Full length human recombit protein of human PDLIM5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.