Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDLIM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113659 Supplier Invitrogen™ Supplier No. PA5113659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNYSVSLVGP APWGFRLQGG KDFNMPLTIS SLKDGGKAAQ ANVRIGDVVL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 86%.

May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDLIM5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PDLIM5
Gene Accession No. Q96HC4
Gene Alias 1110001A05Rik; AI987914; C87059; ENH; Enh1; Enh2; Enh3; Enigma homolog; enigma-like LIM domain protein; enigma-like PDZ and LIM domains protein; L9; LIM; Pdlim5; PDZ and LIM domain 5; PDZ and LIM domain protein 5
Gene Symbols PDLIM5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human PDLIM5 (aa 2-51).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10611
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.